missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
GREENER_CHOICE

Catalog No. SH3098801
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3098801 50 L
SH3098802 100 L
SH3098803 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3098801 Supplier Thermo Scientific™ Supplier No. SH30988.01

Thermo Scientific 3D Tank Liners are composed of Aegis 5-14 film with dimensions optimized for Thermo Scientific drums and commonly-used, industry-standard cylindrical tanks. The tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids

Specifications

Capacity (Metric) 50 L
Quantity 50 L
Tubing Style C-Flex