missing translation for 'onlineSavingsMsg'
Learn More

3B Scientific™ N-Doped Germanium on Printed Circuit Board

Interchangeable board with n-doped germanium crystals to investigate conductivity and Hall potential as function of temperature.

Supplier:  3B Scientific™ U8487030

Catalog No. S08156


Only null left

Description

Description

  • Integrated resistive heating element with temperature sensor directly under crystal, and multi pin plug for connecting circuit board to basic Hall effect apparatus

Specifications:

  • Crystal dimensions: 20 x 10 x 1mm3 approx.
  • Overall dimensions: 70 x 70 x 10mm3 approx.
  • Weight: 30g approx.
Specifications

Specifications

30 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions