Learn More
3B Scientific™ Magnetic Field Lines Demonstrator
Catalog No. S90378
Change view
Click to view available options
Product Type:
Magnetic Field Line Apparatus
Catalog No. Product Type
S90378 Magnetic Field Line Apparatus
1 options
Catalog No. S90378 Supplier 3B Scientific™ Supplier No. U19560
Includes
Magnets, experiment manual
Use with an overhead projector to demonstrate the presence of magnetic field lines.
  • Transparent plastic vessel filled with a liquid containing magnetic powder Dimensions: 11 x 7 x 0.35 in. (280 x 180 x 9mm)
Specifications
For Use With (Application) Overhead projector to demonstrate the presence of magnetic field line
Product Type Magnetic Field Line Apparatus
Includes Magnets, experiment manual
Weight (Metric) 0.82 kg
Weight (English) 1.81 lb.