missing translation for 'onlineSavingsMsg'
Learn More

3A-Amino-3A-deoxy-(2AS,3AS)-alpha-cyclodextrin Hydrate 90.0+%, TCI America™

Catalog No. p-7117743
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2122200MG 200 mg
A21221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2122200MG Supplier TCI America Supplier No. A2122200MG
Only null left

Chemical Identifiers

CAS 121916-94-7
Molecular Formula C36H61NO29
Molecular Weight (g/mol) 971.862
InChI Key TXBHONGFOHSVMN-IIRXRYJYSA-N
PubChem CID 131869226
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)N)O

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 971.86
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3A-Amino-3A-deoxy-(2AS,3AS)-alpha-cyclodextrin Hydrate