missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ 384-Well Skirted Microplates, BIO-CERT™ PCR QUALITY
Optimized for reliable and precise results in a wide range of PCR applications.
$1140.94 - $2457.13
Specifications
| Barcode | No Barcode |
|---|---|
| Binding Property | Untreated |
| Format | Standard |
| Array | 16 x 24 |
| Autoclavable | Autoclavable |
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|
13-882-76
|
BRANDTECH™ BRAND™
781345 |
Clear |
Pack of 50 Each for $1,140.94
|
|
|||||
|
13-878-207
|
BRANDTECH™ BRAND™
781358 |
White |
Pack of 50 Each for $2,457.13
|
|
|||||
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
- Exceptional Quality and Purity
- BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
- Free of contaminants that could interfere with reagents and samples
- Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
- Precision Thermal Performance
- Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
- Designed for rapid and accurate heat transfer, reducing cycle times
- Wide Compatibility
- Suitable for most commercially available thermal cyclers
- White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
- User-Friendly Features
- Blue alphanumeric coding for easy sample identification
- Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
- Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
Specifications
| No Barcode | |
| Standard | |
| Autoclavable | |
| Single-use | |
| Fully skirted | |
| 0.03 mL | |
| Non-sterile | |
| 0.03 mL | |
| Yes |
| Untreated | |
| 16 x 24 | |
| Bagged | |
| Thermal Cyclers | |
| Polypropylene | |
| 384 | |
| Non-treated | |
| Conical Bottom (V-well) |