Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
GREENER_CHOICE
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
$1,264.39 - $2,632.75
Specifications
Format Standard
Array 16 x 24
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
View More Specs
Products 5
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
07000890
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
781900
Black
Case of 40 Each for $1,264.39
Only null left
07000892
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
781906
Black, Clear Bottom
Case of 40 Each for $2,632.75
Only null left
07000891
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
781904
White
Case of 40 Each for $1,308.75
Only null left
07000060
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
781901
Clear
Case of 40 Each for $1,282.00
Only null left
07000031
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
781903
White, Clear Bottom
Case of 40 Each for $2,599.36
Only null left
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Higher assay sensitivity with reduced background
Service Plan
No
Specifications
Specifications
Standard
Polystyrene
Non-sterile
16 x 24
384