missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 384-Well Polypropylene Plates
GREENER_CHOICE

Catalog No. 12565507
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12565507 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12565507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Feature higher well density for economical use of expensive reagents

  • Ideal for storage of compounds and nucleic acid libraries
  • Chemical-resistant
  • Withstand temperatures from -80° to +80°C (-112° to +176°F)
  • Conform to ANSI™-standard footprint for optimal use in automated handling and liquid-dispensing systems

Specifications

Certifications/Compliance ANSI
Format Standard
Array 16 x 24
Color Natural
Product Type 384-Well Storage Microplate
Material Polypropylene
No. of Wells 384
Sterility Non-sterile
For Use With (Application) For storage of compounds and nucleic acid libraries