Learn More
Greiner Bio-One 384-well Polypropylene PCR Microplates
Description
- Free of detectable DNase, RNase, human DNA
- Suitable for real-time, quantitative PCR
- Comprehensive thermal cycler compatibility
- Packaging labeled with product symbols for easier identification and warehousing
Specifications
Specifications
| Format | Standard |
| Description | Roche LightCycler compatible |
| Array | 16 x 24 |
| Color | Natural |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Non-sterile |
For Research use only.