missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well, Fibronectin-Treated, Flat-Bottom, Low Flange Microplate
Description
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.06 cm2 |
| Array | 16 x 24 |
| Packaging | 20/Pk., 100/Cs. |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| Volume (Metric) Well | 110.5 μL |
| No. of Wells | 384 |
| Sterility | Sterile |
| Show More |