Learn More
Corning™ 384 Well Flat Clear Bottom Polystyrene NBS™ Microplates, low flange, no lid, nonsterile
GREENER_CHOICE
Catalog No. 07201014
Change view
Click to view available options
Color:
Black
Catalog No. Color
07201014 Black
1 options
Catalog No. 07201014 Supplier Corning™ Supplier No. 3766
Get Up to $2,500 in Products with Purchase
View all promotions
Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise. Corning™ 384 Well Flat Clear Bottom Polystyrene NBS™ Microplates feature black wall plates that have low background fluorescence, minimal light scatter, and reduced crosstalk.
  • Black wall plates have low background fluorescence, minimal light scatter, and reduced crosstalk
  • Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimized molecular interactions
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
  • Suited for fluorescent assays using either top or bottom detection plate readers
  • Recommended working volume of up to 20 to 80μL
  • Conforms to standard microplate footprint and dimensions
Specifications
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Non-sterile
Surface Treatment Non-binding
Well Shape Square, Flat Bottom (F-well)
No. per Pack 25