missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 384-well, Advanced TC Treated, Flat-Bottom Microplate
Color:White, Clear Bottom, White; Sterility:Sterile; Lid:With Lid
Supplier: Greiner Bio-One 781983
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Non-cytotoxic
- Improved cell adherence
- Permits usage of serum-reduced or serum-free media
- Optimal cultivation conditions for sensitive cells
Specifications
| Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic | |
| With Lid | |
| 16 x 24 | |
| White, Clear Bottom | |
| 384 | |
| Sterile | |
| Rounded Square | |
| 4000 x g (swinging-bucket rotor) | |
| 8 | |
| Yes |
| Standard | |
| 10 mm2 | |
| Not Autoclavable | |
| Polystyrene | |
| 15 to 110 μL | |
| Advanced TC | |
| Yes | |
| 32 | |
| 32/Pk. | |
| 128 x 85 x 15 mm |