missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 382 Microporous Nitrile Coated Nylon Gloves

Catalog No. 296367542
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296367542 2X-Large
296367538 Small
296367539 Medium
296367540 Large
296367541 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 296367542 Supplier SHOWA™ Supplier No. 382XXL10

General purpose gloves designed to prevent fingerprints on metal sheets and glass. Nitrile coating disperses oil away to enhance grip for precision handling in greasy environments.

  • Gloves help to prevent fingerprints on metal sheets and glass
  • Microporous nitrile coating disperses oil away to enhance grip in greasy environments
  • Ergonomic design ensures comfort and breathability
  • Silicone-free

Specifications

Certifications/Compliance CE Category II, EN 388:2016+A1:2018 4121X
Certifications/Compliance (Europe) CE Category II, EN 388:2016+A1:2018 4121X
Construction Seamless Knit
Coating Material Microporous Nitrile
Color Blue, Black
Exterior Finish Grip Pattern
Hand Specific
Latex Free Yes
Material Nylon
Product Type Seamless Knit Gloves
Size 2X-Large
Style Knit
For Use With (Application) Automotive Repairs, and Intricate/Oily Parts Handling
Gender Unisex
Glove Cuff Style Knit
Grip Design Embossed
Series 382
Product Line SHOWA
Show More Show Less