missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 382 Microporous Nitrile Coated Nylon Gloves

Catalog No. 296367540
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296367540 Large
296367538 Small
296367539 Medium
296367541 X-Large
296367542 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 296367540 Supplier SHOWA™ Supplier No. 382L08

General purpose gloves designed to prevent fingerprints on metal sheets and glass. Nitrile coating disperses oil away to enhance grip for precision handling in greasy environments.

  • Gloves help to prevent fingerprints on metal sheets and glass
  • Microporous nitrile coating disperses oil away to enhance grip in greasy environments
  • Ergonomic design ensures comfort and breathability
  • Silicone-free

Specifications

Certifications/Compliance CE Category II, EN 388:2016+A1:2018 4121X
Certifications/Compliance (Europe) CE Category II, EN 388:2016+A1:2018 4121X
Construction Seamless Knit
Coating Material Microporous Nitrile
Color Blue, Black
Exterior Finish Grip Pattern
Hand Specific
Latex Free Yes
Material Nylon
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Automotive Repairs, and Intricate/Oily Parts Handling
Gender Unisex
Glove Cuff Style Knit
Grip Design Embossed
Series 382
Product Line SHOWA
Show More Show Less