Learn More
Celltreat Scientific Products 35mm x 10mm Tissue Culture Treated Dish, Sterile
Supplier:  Celltreat Scientific Products 229635
Encompass
CELLTREAT Scientific Products 35mm x 10mm Tissue Culture Treated Dish. Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 5mL and growth area of 8.5cm. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 500/case.
Catalog No. 50202030