Learn More
Mattek Corporation 35MM GLS BTM PETRI DISH PK/10
Supplier:  Mattek Corporation P35G1.514C/SLEEVE
Encompass
Culture Dish; Glass Bottom; Thickness: No. 1.5; Uncoated; Dish dia.: 35mm; Microwell dia.: 14mm. This item is a package size of 10. Item is also available in a case of 200. Case size does have pricing break. See NC9268399 for case size.
Catalog No. NC9341562
Only null left