Learn More
Cem Corporation 35MIL SPD PLUS VIAL CAP 25/PK
Supplier: Cem Corporation 909350
35mL/80mL SP-D Plus Pressure Caps. Includes quantity 25 of 35mL/80mL vial caps with TFM septa. Compatible with Discover SP-D.