missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ 33 Gallon Yellow Infectious Linen Bags

Catalog No. 17988502
Change view
Click to view available options
Capacity (English):
33 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17988502 33 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17988502 Supplier Eagle™ Supplier No. 1692
Only null left

Yellow infectious linen bags can be used to minimize waste by segregating infectious linens for laundering.

  • Designed to fit Eagle™ 1655 (Nestable) and 1656 (Straight sided) series drums
  • Seal bag in place and close drum using its lid and lever-lock once full and ready for laundering
  • Yellow color bag is easily identifiable for containing infectious linens
  • Drums can be yellow, black, or blue
  • English, Spanish and French translations along with international recognizable biohazard symbol
  • High level of flexibility and shear resistance can be attained without leaks
  • Tubular construction, single side seal and star seal bottom provide stable, leak proof bag
  • Meets ASTM D 1709-01 and ASTM D 1922-00a specifications
  • Linear low density polyethylene (LLDPE)
  • Thickness: 1.2 mil

Specifications

Capacity (English) 33 gal.
Color Yellow
Quantity 100/Pk.
Autoclavable Not Autoclavable
Liner Material LDPE
For Use With (Equipment) Eagle™ Drums
Width (English) 32.5 in.
Length (English) 45 in.
Thickness 1.2 mil
Height (English) 32.5 in.
Material Polyethylene
Product Type Biohazard Linen Bags
Show More Show Less