missing translation for 'onlineSavingsMsg'
Learn More

3-Undecanol 94.0+%, TCI America™

Catalog No. U00285ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00285ML 5 mL
U002825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00285ML Supplier TCI America Supplier No. U00285ML
Only null left

Chemical Identifiers

CAS 6929-08-4
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00046730
InChI Key HCARCYFXWDRVBZ-UHFFFAOYNA-N
PubChem CID 98970
IUPAC Name undecan-3-ol
SMILES CCCCCCCCC(CC)O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 mL
UN Number 1987
Formula Weight 172.31
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Undecanol
EINECSNumber (2)-0217&(2)-3704
TSCA No