missing translation for 'onlineSavingsMsg'
Learn More

3-[Tris(trimethylsilyloxy)silyl]propyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. p-7132661
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M132425ML 25 mL
M1324100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M132425ML Supplier TCI America Supplier No. M132425ML

Chemical Identifiers

CAS 17096-07-0
Molecular Formula C16H38O5Si4
Molecular Weight (g/mol) 422.815
MDL Number MFCD00053871
InChI Key BESKSSIEODQWBP-UHFFFAOYSA-N
Synonym 3-methacryloyloxy propyltris trimethylsiloxy silane, 3-tris trimethylsiloxy silyl propyl methacrylate, 3-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxan-3-yl propyl methacrylate, unii-d32s78j7t8, 3-methacryloyloxypropyl tris trimethylsiloxy silane, 3-methacryloxypropyltris trimethylsiloxy silane, 3-tris trimethylsilyloxy silyl propyl methacrylate, methacryloxypropyltris trimethylsiloxy silane, 3-3,3,3-trimethyl-1,1-bis trimethylsilyl oxy disiloxanyl propyl methacrylate
PubChem CID 123371
IUPAC Name 3-tris(trimethylsilyloxy)silylpropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 115°C
Quantity 25 mL
Formula Weight 422.82
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-[Tris(trimethylsilyloxy)silyl]propyl Methacrylate (stabilized with MEHQ)
EINECSNumber (7)-0450
TSCA No