missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[Tris(trimethylsilyloxy)silyl]propyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 115°C |
| Quantity | 100 mL |
| Formula Weight | 422.82 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-[Tris(trimethylsilyloxy)silyl]propyl Methacrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (7)-0450 |
| TSCA | No |