missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilylethynyl)pyridine 98.0+%, TCI America™

Catalog No. T19631G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19631G Supplier TCI America Supplier No. T19631G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80673-00-3
Molecular Formula C10H13NSi
Molecular Weight (g/mol) 175.306
MDL Number MFCD01863744
InChI Key MTMBLPOPETZZRX-UHFFFAOYSA-N
Synonym 3-trimethylsilyl ethynyl pyridine, 3-trimethylsilylethynyl pyridine, 3-pyridylethynyl trimethylsilane, 3-2-trimethylsilyl ethynyl pyridine, trimethyl 2-pyridin-3-ylethynyl silane, acmc-209pjw, 3-trimethylsilylethynylpyridine, trimethyl-2-3-pyridyl ethynyl silane, 1-3-pyridyl-2-trimethylsilylacetylene
PubChem CID 4100857
IUPAC Name trimethyl(2-pyridin-3-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CN=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 175.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trimethylsilylethynyl)pyridine
TSCA No
Recommended Storage Freezer