missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propiolic Acid 97.0+%, TCI America™

Catalog No. T30405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30405G 5 g
T30401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30405G Supplier TCI America Supplier No. T30405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5683-31-8
Molecular Formula C6H10O2Si
Molecular Weight (g/mol) 142.23
MDL Number MFCD00190216
InChI Key IPEATTYBFBRNEB-UHFFFAOYSA-N
Synonym 3-trimethylsilyl propiolic acid, 3-trimethylsilyl propynoic acid, trimethylsilylpropiolic acid, 3-trimethylsilylpropynoic acid, 3-trimethylsilyl prop-2-ynoic acid, 2-propynoic acid, 3-trimethylsilyl, tms-propynoic acid, acmc-209lul, 3-trimethylsilyl propynoicacid, 3-trimethylsilyl propalic acid
PubChem CID 2760863
IUPAC Name 3-(trimethylsilyl)prop-2-ynoic acid
SMILES C[Si](C)(C)C#CC(O)=O

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 142.23
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trimethylsilyl)propiolic Acid
TSCA No
Recommended Storage Refrigerator