missing translation for 'onlineSavingsMsg'
Learn More

3-[(Trimethylsilyl)ethynyl]pyridine, 97%

Catalog No. p-7052639
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5510103 1 g
AAH5510106 5 g
AAH5510114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5510103 Supplier Thermo Scientific Chemicals Supplier No. H5510103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80673-00-3 | C10H13NSi | 175.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80673-00-3
Molecular Formula C10H13NSi
Molecular Weight (g/mol) 175.306
MDL Number MFCD01863744
InChI Key MTMBLPOPETZZRX-UHFFFAOYSA-N
Synonym 3-trimethylsilyl ethynyl pyridine, 3-trimethylsilylethynyl pyridine, 3-pyridylethynyl trimethylsilane, 3-2-trimethylsilyl ethynyl pyridine, trimethyl 2-pyridin-3-ylethynyl silane, acmc-209pjw, 3-trimethylsilylethynylpyridine, trimethyl-2-3-pyridyl ethynyl silane, 1-3-pyridyl-2-trimethylsilylacetylene
PubChem CID 4100857
IUPAC Name trimethyl(2-pyridin-3-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CN=CC=C1

Specifications

Color Colorless to Yellow
Density 0.922 g/mL
Boiling Point 229°C
Flash Point 94°C (201°F)
Refractive Index 1.529
Quantity 1 g
Formula Weight 175.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-[(Trimethylsilyl)ethynyl]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only