missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)pyrazole 98.0+%, TCI America™

Catalog No. T27655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27655G 5 g
T27651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27655G Supplier TCI America Supplier No. T27655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20154-03-4
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.077
MDL Number MFCD00115018
InChI Key PYXNITNKYBLBMW-UHFFFAOYSA-N
Synonym 3-trifluoromethyl pyrazole, 3-trifluoromethyl-1h-pyrazole, 5-trifluoromethyl-1h-pyrazole, 3-trifluoromethylpyrazole, trifluoromethylpyrazole, 1h-pyrazole, 3-trifluoromethyl, 3-trifluoromethyl-2h-pyrazole, pubchem10164, pubchem10423
PubChem CID 1807034
IUPAC Name 5-(trifluoromethyl)-1H-pyrazole
SMILES C1=C(NN=C1)C(F)(F)F

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 98°C
Quantity 5 g
Formula Weight 136.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)pyrazole