missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-3-(Trifluoromethyl)piperidine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 768-31-0 |
|---|---|
| Molecular Formula | C6H11F3N |
| Molecular Weight (g/mol) | 154.16 |
| MDL Number | MFCD02183561 |
| InChI Key | JOHFJTBDUSVGQB-RXMQYKEDSA-O |
| Synonym | 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl |
| PubChem CID | 2760776 |
| IUPAC Name | 3-(trifluoromethyl)piperidine |
| SMILES | FC(F)(F)[C@@H]1CCC[NH2+]C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC454820010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C6H11F3N | |
| MFCD02183561 | |
| 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl | |
| 3-(trifluoromethyl)piperidine |
Specifications
| 768-31-0 | |
| 28°C | |
| 97.5% min. (GC) | |
| MFCD02183561 | |
| JOHFJTBDUSVGQB-RXMQYKEDSA-O | |
| 3-(trifluoromethyl)piperidine | |
| 2760776 | |
| 98% |
| 128.0°C to 130.0°C | |
| Authentic | |
| C6H11F3N | |
| 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl | |
| FC(F)(F)[C@@H]1CCC[NH2+]C1 | |
| 154.16 | |
| 153.15 | |
| (±)-3-(Trifluoromethyl)piperidine |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
RUO – Research Use Only