missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)piperidine, 97%

Catalog No. AAH25851MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25851MD 250 mg
AAH2585103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25851MD Supplier Thermo Scientific Chemicals Supplier No. H25851MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 768-31-0 | C6H11F3N | 154.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 768-31-0
Molecular Formula C6H11F3N
Molecular Weight (g/mol) 154.16
MDL Number MFCD02183561
InChI Key JOHFJTBDUSVGQB-RXMQYKEDSA-O
PubChem CID 2760776
SMILES FC(F)(F)[C@@H]1CCC[NH2+]C1

Specifications

Density 1.16
Boiling Point 128°C to 130°C
Flash Point 28°C (83°F)
Odor Amine-like
Refractive Index 1.393
Quantity 250 mg
UN Number UN2733
Sensitivity Air Sensitive
Synonym 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl
IUPAC Name (3R)-3-(trifluoromethyl)piperidin-1-ium
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)piperidine
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only