missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)cyclohexanone, 97%, Thermo Scientific Chemicals

Catalog No. AAH3408703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3408703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3408703 Supplier Thermo Scientific Chemicals Supplier No. H3408703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 585-36-4 | Molecular Formula: C7H9F3O | Molecular Weight: 166.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 585-36-4
Molecular Formula C7H9F3O
Molecular Weight (g/mol) 166.14
MDL Number MFCD00034662
InChI Key YHADNJOEQWBXAR-UHFFFAOYNA-N
Synonym 3-trifluoromethyl cyclohexanone, 3-trifluoromethyl cyclohexan-1-one, cyclohexanone, 3-trifluoromethyl, 3-trifluoromethyl-cyclohexanone, 3-trifluoromethyl-1-cyclohexanone, 1-oxo-3-trifluoromethyl cyclohexane
PubChem CID 2779355
IUPAC Name 3-(trifluoromethyl)cyclohexan-1-one
SMILES FC(F)(F)C1CCCC(=O)C1

Specifications

Boiling Point 82°C (29 mmHg)
Quantity 1 g
UN Number UN1224
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)cyclohexanone
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only