missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)cinnamic Acid 99.0+%, TCI America™

Catalog No. T15475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15475G 5 g
T154725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15475G Supplier TCI America Supplier No. T15475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 779-89-5
Molecular Formula C10H7F3O2
Molecular Weight (g/mol) 216.159
MDL Number MFCD00004393
InChI Key KSBWHDDGWSYETA-SNAWJCMRSA-N
Synonym 3-trifluoromethyl cinnamic acid, 3-3-trifluoromethyl phenyl acrylic acid, m-trifluoromethyl cinnamic acid, e-3-trifluoromethyl cinnamic acid, unii-p9lj9on37w, 2e-3-3-trifluoromethyl phenyl prop-2-enoic acid, e-m-trifluoromethyl cinnamic acid, 3-e-trifluoromethyl cinnamic acid
PubChem CID 719451
ChEBI CHEBI:60704
IUPAC Name (E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)C=CC(=O)O

Specifications

Melting Point 135°C
Color White
Quantity 5 g
Formula Weight 216.16
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)cinnamic Acid