missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)cinnamic acid, predominantly trans, 98+%

Catalog No. AAA1698314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1698314 25 g
AAA1698306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1698314 Supplier Thermo Scientific Chemicals Supplier No. A1698314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 779-89-5 | C10H7F3O2 | 216.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 779-89-5
Molecular Formula C10H7F3O2
Molecular Weight (g/mol) 216.159
MDL Number MFCD00004393
InChI Key KSBWHDDGWSYETA-SNAWJCMRSA-N
Synonym 3-trifluoromethyl cinnamic acid, 3-3-trifluoromethyl phenyl acrylic acid, m-trifluoromethyl cinnamic acid, e-3-trifluoromethyl cinnamic acid, unii-p9lj9on37w, 2e-3-3-trifluoromethyl phenyl prop-2-enoic acid, e-m-trifluoromethyl cinnamic acid, 3-e-trifluoromethyl cinnamic acid
PubChem CID 719451
ChEBI CHEBI:60704
IUPAC Name (E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)C=CC(=O)O

Specifications

Melting Point 132°C to 136°C
Odor Odorless
Quantity 25 g
Beilstein 1963372
Formula Weight 216.16
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)cinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-301-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only