missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific™

Catalog No. AC312940050
Change view
Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312940050 5mL Glass bottle
AC312940250 25mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312940050 Supplier Thermo Scientific Chemicals Supplier No. 312940050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2251-65-2
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.57
MDL Number MFCD00000680
InChI Key RUJYJCANMOTJMO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-trifluoromethyl, m-trifluoromethyl benzoyl chloride, m-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-m-toluoyl chloride, 3-trifluoromethyl benzoylchloride, 3-trifluoromethyl-benzoyl chloride, alpha,alpha,alpha-trifluoro-m-tolyl chloride, pubchem5468
PubChem CID 75257
IUPAC Name 3-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.3800g/mL
Boiling Point 184.0°C to 186.0°C (750.0mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780
Linear Formula CF3C6H4COCl
Quantity 5mL
Specific Gravity 1.38
Solubility Information Solubility in water: decomposes
Formula Weight 208.57
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-844-5

RUO – Research Use Only