missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethyl)acetophenone 97.0+%, TCI America™

$34.66 - $132.84

Chemical Identifiers

CAS 349-76-8
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000391
InChI Key ABXGMGUHGLQMAW-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl acetophenone, 3-trifluoromethyl acetophenone, 3-trifluoromethylacetophenone, ethanone, 1-3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl ethanone, m-trifluoromethylacetophenone, 3'-trifluoromethylacetophenone, 1-3-trifluoromethyl phenyl ethan-1-one, 1-3-trifluoromethyl-phenyl-ethanone
PubChem CID 67682
IUPAC Name 1-[3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T15465G
View Documents
TCI America
T15465G
5 g
Each for $34.66
Only null left
 
T154625G
View Documents
TCI America
T154625G
25 g
Each for $132.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

349-76-8
188.149
ABXGMGUHGLQMAW-UHFFFAOYSA-N
67682
CC(=O)C1=CC(=CC=C1)C(F)(F)F
C9H7F3O
MFCD00000391
3'-trifluoromethyl acetophenone, 3-trifluoromethyl acetophenone, 3-trifluoromethylacetophenone, ethanone, 1-3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl ethanone, m-trifluoromethylacetophenone, 3'-trifluoromethylacetophenone, 1-3-trifluoromethyl phenyl ethan-1-one, 1-3-trifluoromethyl-phenyl-ethanone
1-[3-(trifluoromethyl)phenyl]ethanone

Specifications

349-76-8
C9H7F3O
5 g
ABXGMGUHGLQMAW-UHFFFAOYSA-N
1-[3-(trifluoromethyl)phenyl]ethanone
67682
≥97.0% (GC)
3′-(Trifluoromethyl)acetophenone
199°C
MFCD00000391
3'-trifluoromethyl acetophenone, 3-trifluoromethyl acetophenone, 3-trifluoromethylacetophenone, ethanone, 1-3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl ethanone, m-trifluoromethylacetophenone, 3'-trifluoromethylacetophenone, 1-3-trifluoromethyl phenyl ethan-1-one, 1-3-trifluoromethyl-phenyl-ethanone
CC(=O)C1=CC(=CC=C1)C(F)(F)F
188.149
188.15
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents