missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethyl)acetophenone, 98+%

Catalog No. p-7081987
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1221006 5 g
AAA1221014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1221006 Supplier Thermo Scientific Chemicals Supplier No. A1221006

CAS: 349-76-8 | C9H7F3O | 188.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 349-76-8
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000391
InChI Key ABXGMGUHGLQMAW-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl acetophenone, 3-trifluoromethyl acetophenone, 3-trifluoromethylacetophenone, ethanone, 1-3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl ethanone, m-trifluoromethylacetophenone, 3'-trifluoromethylacetophenone, 1-3-trifluoromethyl phenyl ethan-1-one, 1-3-trifluoromethyl-phenyl-ethanone
PubChem CID 67682
IUPAC Name 1-[3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Density 1.26
Boiling Point 198°C to 200°C
Flash Point 83°C (181°F)
Odor Characteristic
Refractive Index 1.4635
Quantity 5 g
Beilstein 640151
Formula Weight 188.15
Percent Purity ≥98%
Chemical Name or Material 3'- (Trifluoromethyl)acetophenone
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-490-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only