missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethoxyphenylboronic acid, 98%, Thermo Scientific™

Catalog No. AC359720010
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC359720010 1g
AC359720050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC359720010 Supplier Thermo Scientific Chemicals Supplier No. 359720010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 179113-90-7
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
InChI Key UWDFWVLAHRQSKK-UHFFFAOYSA-N
Synonym 3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl
PubChem CID 2734385
IUPAC Name [3-(trifluoromethoxy)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)OC(F)(F)F)(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 205.93
Percent Purity 98%
Chemical Name or Material 3-Trifluoromethoxyphenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only