missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T23625G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23625G Supplier TCI America Supplier No. T23625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 179113-90-7
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320697
InChI Key UWDFWVLAHRQSKK-UHFFFAOYSA-N
Synonym 3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl
PubChem CID 2734385
IUPAC Name [3-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC(OC(F)(F)F)=C1

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Formula Weight 205.93
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethoxy)phenylboronic Acid (contains varying amounts of Anhydride)