missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7137164
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23341G 1 g
T23345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23341G Supplier TCI America Supplier No. T23341G

Chemical Identifiers

CAS 86270-03-3
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD00061268
InChI Key MGKIOAXLPYJSMU-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzoyl chloride, m-trifluoromethoxybenzoyl chloride, m-trifluoromethoxy benzoyl chloride, benzoyl chloride, 3-trifluoromethoxy, 3-trifluoromethoxy-benzoyl chloride, benzoylchloride, 3-trifluoromethoxy, ksc450s6d, m-trifluoromethoxybenzoylchloride, 3-trifluoromethoxy benzoylchloride, 3-trifluoromethyloxy benzoyl chloride
PubChem CID 522854
IUPAC Name 3-(trifluoromethoxy)benzoyl chloride
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C(=O)Cl

Specifications

Color Colorless
Quantity 1 g
UN Number 3265
Formula Weight 224.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)benzoyl Chloride