missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzeneboronic acid, 98%

CAS: 179113-90-7 | C7H6BF3O3 | 205.93 g/mol

Supplier:  Thermo Scientific Chemicals L1977506

 View more versions of this product

Catalog No. AAL1977506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Trifluoromethoxy)benzeneboronic acid
∼80°C
MFCD01320697
7531456
UWDFWVLAHRQSKK-UHFFFAOYSA-N
[3-(trifluoromethoxy)phenyl]boronic acid
2734385
98%
179113-90-7
C7H6BF3O3
5 g
3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl
OB(O)C1=CC=CC(OC(F)(F)F)=C1
205.93
205.93

Chemical Identifiers

179113-90-7
205.93
UWDFWVLAHRQSKK-UHFFFAOYSA-N
2734385
OB(O)C1=CC=CC(OC(F)(F)F)=C1
C7H6BF3O3
MFCD01320697
3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl
[3-(trifluoromethoxy)phenyl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only