missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzeneboronic acid, 98%

Catalog No. AAL1977503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1977503 1 g
AAL1977506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1977503 Supplier Thermo Scientific Chemicals Supplier No. L1977503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 179113-90-7 | C7H6BF3O3 | 205.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 179113-90-7
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320697
InChI Key UWDFWVLAHRQSKK-UHFFFAOYSA-N
Synonym 3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl
PubChem CID 2734385
IUPAC Name [3-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC(OC(F)(F)F)=C1

Specifications

Melting Point ∼80°C
Quantity 1 g
Beilstein 7531456
Formula Weight 205.93
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only