missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoroacetyl)pyridine, 95%

Catalog No. p-7056962
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66214MD 250 mg
AAH6621403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66214MD Supplier Thermo Scientific Chemicals Supplier No. H66214MD
Only null left

CAS: 33284-21-8 | C7H6F3NO | 177.126 g/mol

3-(Trifluoroacetyl)pyridine is used as medical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoroacetyl)pyridine is used as medical intermediates.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 33284-21-8
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD16067922
InChI Key LHXKPHQPUHHFQW-UHFFFAOYSA-N
Synonym 2,2,2-trifluoro-1-pyridin-3-yl ethanol, 2,2,2-trifluoro-1-pyridin-3-yl ethan-1-ol, 2,2,2-trifluoro-1-3-pyridyl ethanol, alpha-trifluoromethyl pyridine-3-methanol, 10.14272/lhxkphqpuhhfqw-uhfffaoysa-n, 3-2,2,2-trifluoro-1-hydroxyethyl pyridine, doi:10.14272/lhxkphqpuhhfqw-uhfffaoysa-n
PubChem CID 11217557
IUPAC Name 2,2,2-trifluoro-1-pyridin-3-ylethanol
SMILES C1=CC(=CN=C1)C(C(F)(F)F)O

Specifications

Quantity 250 mg
Beilstein 4800857
Formula Weight 175.11
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(Trifluoroacetyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only