missing translation for 'onlineSavingsMsg'
Learn More

3-(Triallylsilyl)propyl Acrylate (stabilized with MEHQ) 92.0+%, TCI America™

Catalog No. t32285g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32285G Supplier TCI America Supplier No. T32285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1990509-29-9
Molecular Formula C15H24O2Si
Synonym Acrylic Acid 3-(Triallylsilyl)propyl Ester

Specifications

Quantity 5 g
Formula Weight 264.44
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Triallylsilyl)propyl Acrylate (stabilized with MEHQ)