missing translation for 'onlineSavingsMsg'
Learn More

3-Tolylboronic acid, 97%

Catalog No. p-201586
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC344420010 1 g Glass bottle
AC344420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC344420010 Supplier Thermo Scientific Chemicals Supplier No. 344420010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17933-03-8 | C7H9BO2 | 135.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17933-03-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00040198
InChI Key BJQCPCFFYBKRLM-UHFFFAOYSA-N
Synonym 3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
PubChem CID 2733950
IUPAC Name (3-methylphenyl)boronic acid
SMILES CC1=CC=CC(=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160.0°C to 162.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C6H4B(OH)2
Quantity 1 g
Solubility Information Solubility in water: +/- 2.5% in water.
Formula Weight 135.96
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 3-Tolylboronic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
TSCA TSCA

RUO – Research Use Only