missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiophenemethylamine 97.0+%, TCI America™
Supplier: TCI America T28501G
Specifications
| 3-Thiophenemethylamine | |
| Yellow | |
| MFCD01529872 | |
| 2735 | |
| DUDAKCCDHRNMDJ-UHFFFAOYSA-N | |
| thiophen-3-ylmethanamine | |
| 2776381 | |
| ≥97.0% (GC,T) |
| 27757-86-4 | |
| C5H7NS | |
| 1 g | |
| 3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine | |
| C1=CSC=C1CN | |
| 113.178 | |
| 113.18 | |
| Liquid |
Chemical Identifiers
| 27757-86-4 | |
| 113.178 | |
| DUDAKCCDHRNMDJ-UHFFFAOYSA-N | |
| 2776381 | |
| C1=CSC=C1CN |
| C5H7NS | |
| MFCD01529872 | |
| 3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine | |
| thiophen-3-ylmethanamine |
Safety and Handling
TSCA : No