missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenemethylamine, 96%

Catalog No. AAH6148706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6148706 5 g
AAH6148703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6148706 Supplier Thermo Scientific Chemicals Supplier No. H6148706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27757-86-4 | C5H7NS | 113.178 g/mol

3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store in cool, dry place in tightly closed container. With good ventilation. Store away from air.

Chemical Identifiers

CAS 27757-86-4
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD01529872
InChI Key DUDAKCCDHRNMDJ-UHFFFAOYSA-N
Synonym 3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine
PubChem CID 2776381
IUPAC Name thiophen-3-ylmethanamine
SMILES C1=CSC=C1CN

Specifications

Density 1.13
Boiling Point 102°C (15 mmHg)
Refractive Index 1.566
Quantity 5 g
UN Number UN2735
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 113.18
Percent Purity 97%
Chemical Name or Material 3-Thiophenemethylamine
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only