missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenemethanol 96.0+%, TCI America™

Catalog No. p-7136894
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16605G 5 g
T166025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16605G Supplier TCI America Supplier No. T16605G
Only null left

Chemical Identifiers

CAS 71637-34-8
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00014534
InChI Key BOWIFWCBNWWZOG-UHFFFAOYSA-N
Synonym 3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
PubChem CID 123570
IUPAC Name (thiophen-3-yl)methanol
SMILES OCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 73°C
Quantity 5 g
Formula Weight 114.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Thiophenemethanol