missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenemethanol, 97%

Catalog No. p-7043659
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0193506 5 g
AAL0193514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0193506 Supplier Thermo Scientific Chemicals Supplier No. L0193506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71637-34-8 | C5H6OS | 114.16 g/mol

3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 71637-34-8
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00014534
InChI Key BOWIFWCBNWWZOG-UHFFFAOYSA-N
Synonym 3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
PubChem CID 123570
SMILES OCC1=CSC=C1

Specifications

Density 1.21
Boiling Point 86°C to 88°C (10 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.565
Quantity 5 g
Beilstein 106460
Solubility Information Slightly soluble in water.
IUPAC Name thiophen-3-ylmethanol
Formula Weight 114.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Thiophenemethanol
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 275-741-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only