missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenemalonic Acid 98.0+%, TCI America™

Catalog No. T13545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13545G 5 g
T135425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13545G Supplier TCI America Supplier No. T13545G

Chemical Identifiers

CAS 21080-92-2
Molecular Formula C7H4O4S
Molecular Weight (g/mol) 184.17
MDL Number MFCD00005469
InChI Key GCOOGCQWQFRJEK-UHFFFAOYSA-L
Synonym 3-thiophenemalonic acid, 3-thienylmalonic acid, 2-thiophen-3-yl malonic acid, propanedioic acid, 3-thienyl, 3-thiophene malonic acid, 3-thienylpropanedioic acid, 3-thienyl malonic acid, thiophene-3-malonic acid, 2-3-thienyl malonic acid, 2-3-thienyl propanedioic acid
PubChem CID 88782
IUPAC Name 2-(thiophen-3-yl)propanedioate
SMILES [O-]C(=O)C(C([O-])=O)C1=CSC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 186.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Thiophenemalonic Acid