missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenecarboxaldehyde, 98%, Thermo Scientific™

$130.44 - $1,065.25

Chemical Identifiers

CAS 498-62-4
Molecular Formula C5H4OS
Molecular Weight (g/mol) 112.15
MDL Number MFCD00005466
InChI Key RBIGKSZIQCTIJF-UHFFFAOYSA-N
Synonym 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde
PubChem CID 68135
ChEBI CHEBI:87611
IUPAC Name thiophene-3-carbaldehyde
SMILES C1=CSC=C1C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC187980050
View Documents Promotion Details
N/A 5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC187981000
View Documents Promotion Details
N/A 100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

498-62-4
112.15
RBIGKSZIQCTIJF-UHFFFAOYSA-N
68135
thiophene-3-carbaldehyde
C5H4OS
MFCD00005466
3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde
CHEBI:87611
C1=CSC=C1C=O

Specifications

498-62-4
100.0
194.0°C to 196.0°C
Authentic
C5H4OS
MFCD00005466
1.28
RBIGKSZIQCTIJF-UHFFFAOYSA-N
thiophene-3-carbaldehyde
68135
112.15
3-Thiophenecarboxaldehyde, 98%
97.5
1.2800g/mL
64°C
97.5% min. (GC)
1.5820 to 1.5860
17,III,4497
3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde
C1=CSC=C1C=O
112.15
CHEBI:87611
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protectio

GHS Signal Word: Warning

EINECSNumber : 207-865-5

TSCA : TSCA

Recommended Storage : Product may darken on standing

SDS
Documents

Documents

RUO – Research Use Only