missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneboronic acid, 97%

Catalog No. p-199840
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC345660010 1 g Glass bottle
AC345660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC345660010 Supplier Thermo Scientific Chemicals Supplier No. 345660010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6165-69-1 | C4H5BO2S | 127.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6165-69-1
Molecular Formula C4H5BO2S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00151851
InChI Key QNMBSXGYAQZCTN-UHFFFAOYSA-N
Synonym 3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
PubChem CID 581760
IUPAC Name thiophen-3-ylboronic acid
SMILES OB(O)C1=CSC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 162°C to 167°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 127.96
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Thiopheneboronic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only