missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneacetonitrile, 98%

CAS: 13781-53-8 | C6H5NS | 123.17 g/mol

Supplier:  Thermo Scientific Chemicals A1280514

 View more versions of this product

Catalog No. AAA1280514


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Thiopheneacetonitrile
1.08
119°C (246°F)
1.543
25 g
107774
GWZCLMWEJWPFFA-UHFFFAOYSA-N
2-thiophen-3-ylacetonitrile
83730
98%
13781-53-8
240°C to 242°C
C6H5NS
MFCD00005471
UN3276
3-thiopheneacetonitrile, thiophene-3-acetonitrile, 2-thiophen-3-yl acetonitrile, 3-thienylacetonitrile, 3-cyanomethylthiophene, 3-thiophene acetonitrile, thien-3-ylacetonitrile, 2-3-thienyl acetonitrile, thiophen-3-acetonitrile, 2-3-thienyl ethanenitrile
N#CCC1=CSC=C1
123.17
123.18

Chemical Identifiers

13781-53-8
123.17
GWZCLMWEJWPFFA-UHFFFAOYSA-N
83730
N#CCC1=CSC=C1
C6H5NS
MFCD00005471
3-thiopheneacetonitrile, thiophene-3-acetonitrile, 2-thiophen-3-yl acetonitrile, 3-thienylacetonitrile, 3-cyanomethylthiophene, 3-thiophene acetonitrile, thien-3-ylacetonitrile, 2-3-thienyl acetonitrile, thiophen-3-acetonitrile, 2-3-thienyl ethanenitrile
2-thiophen-3-ylacetonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 237-433-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only