missing translation for 'onlineSavingsMsg'
Learn More

3-Thienylmethylamine, 97%, Thermo Scientific™

Catalog No. p-2700339
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13113DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC13113DE Supplier Thermo Scientific Chemicals Supplier No. CC13113DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27757-86-4
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD01529872
InChI Key DUDAKCCDHRNMDJ-UHFFFAOYSA-N
Synonym 3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine
PubChem CID 2776381
IUPAC Name thiophen-3-ylmethanamine
SMILES C1=CSC=C1CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 113.18
Percent Purity 97%
Chemical Name or Material 3-Thienylmethylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.