missing translation for 'onlineSavingsMsg'
Learn More

(3-Thien-2-ylphenyl)methanol, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC44509DA

 View more versions of this product

Catalog No. CC44509DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

(3-Thien-2-ylphenyl)methanol
96.5
97%
C11H10OS
1g
SATFADRDDGOLJP-UHFFFAOYSA-N
(3-thiophen-2-ylphenyl)methanol
7537586
97%
103669-00-7
100.0
Amber glass bottle
MFCD06203108
3-thien-2-ylphenyl methanol, 3-2-thienyl benzyl alcohol, 3-thiophen-2-yl phenyl methanol, 3-2-thienyl phenyl methanol, benzenemethanol,3-2-thienyl, acmc-1c9kv, 3-thiophen-2-ylphenyl methanol, 3-thiophen-2-yl benzyl alcohol, 3-2-thienyl phenyl methan-1-ol
OCC1=CC=CC(=C1)C1=CC=CS1
190.26
190.27

Chemical Identifiers

103669-00-7
190.26
SATFADRDDGOLJP-UHFFFAOYSA-N
7537586
OCC1=CC=CC(=C1)C1=CC=CS1
C11H10OS
MFCD06203108
3-thien-2-ylphenyl methanol, 3-2-thienyl benzyl alcohol, 3-thiophen-2-yl phenyl methanol, 3-2-thienyl phenyl methanol, benzenemethanol,3-2-thienyl, acmc-1c9kv, 3-thiophen-2-ylphenyl methanol, 3-thiophen-2-yl benzyl alcohol, 3-2-thienyl phenyl methan-1-ol
(3-thiophen-2-ylphenyl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions